Learn More
Apexbio Technology LLC Apramycin Sulfate 65710-07-8 100mg
SDP
Catalog No. 5000091266
Encompass_Preferred
Apramycin Sulfate (65710-07-8) is an aminoglycoside antibiotic targeting bacterial ribosomal RNA It is designed to bind to the ribosomal RNA s conserved deep groove region thereby inhibiting bacterial protein synthesis Apramycin Sulfate exerts its biological activity primarily through specific interaction with bacterial ribosomal RNA leading to inhibition of protein synthesis and disruption of bacterial cell growth In experimental assays Apramycin Sulfate demonstrates inhibitory activity against various bacterial strains with reported IC50 values ranging from 1 to 10 g/mL depending on the strain and experimental conditions Based on these pharmacological properties Apramycin Sulfate holds research potential in antibacterial mechanism studies ribosomal structure-function analyses and resistance mechanism research
Catalog No. 50-000-91266 Supplier Apexbio Technology LLC Supplier No. B1665100
Apramycin Sulfate (65710-07-8) is an aminoglycoside antibiotic targeting bacterial ribosomal RNA It is designed to bind to the ribosomal RNA s conserved deep groove region thereby inhibiting bacterial protein synthesis Apramycin Sulfate exerts its biological activity primarily through specific interaction with bacterial ribosomal RNA leading to inhibition of protein synthesis and disruption of bacterial cell growth In experimental assays Apramycin Sulfate demonstrates inhibitory activity against various bacterial strains with reported IC50 values ranging from 1 to 10 g/mL depending on the strain and experimental conditions Based on these pharmacological properties Apramycin Sulfate holds research potential in antibacterial mechanism studies ribosomal structure-function analyses and resistance mechanism research