Learn More
Medchemexpress LLC Apramycin sulfate | 65710-07-8 | 98.0% | 1 ML
SDP
Catalog No. 5000412063
Encompass_Preferred
Apramycin sulfate is an orally active aminoglycoside antibiotic that demonstrates tolerance to acidic pH conditions and resistance to aminoglycoside-modifying enzymes. This antibiotic functions by targeting bacterial ribosomes, thereby inhibiting protein biosynthesis, and is recognized as a potential agent against drug resistance.
  • Is orally active.
  • Is acidic pH tolerant.
  • Is aminoglycoside-modifying-enzymes-tolerant.
  • Inhibits protein biosynthesis.
  • Targets the bacterial ribosome.
  • Is a potential anti-drug-resistance antibiotic.
Catalog No. 50-004-12063 Supplier Medchemexpress LLC Supplier No. HYB132910MM/1ML
Apramycin sulfate is an orally active aminoglycoside antibiotic that demonstrates tolerance to acidic pH conditions and resistance to aminoglycoside-modifying enzymes. This antibiotic functions by targeting bacterial ribosomes, thereby inhibiting protein biosynthesis, and is recognized as a potential agent against drug resistance.
  • Is orally active.
  • Is acidic pH tolerant.
  • Is aminoglycoside-modifying-enzymes-tolerant.
  • Inhibits protein biosynthesis.
  • Targets the bacterial ribosome.
  • Is a potential anti-drug-resistance antibiotic.