Learn More
Medchemexpress LLC Apramycin sulfate | 65710-07-8 | 98.0% | 500 MG
SDP
Catalog No. 5000413215
Encompass_Preferred
Apramycin sulfate is an orally active, acidic pH tolerant aminoglycoside antibiotic. It inhibits protein biosynthesis by targeting bacterial ribosomes and is considered a potential anti-drug-resistance antibiotic. This compound demonstrates significant efficacy against various bacterial strains, including *K. pneumoniae*, *Enterobacter*, and *E. coli*, often showing better antimicrobial activity than other common antibiotics.
  • Orally active and acidic pH tolerant.
  • Tolerant to aminoglycoside-modifying enzymes.
  • Inhibits protein biosynthesis in bacteria.
  • Potential anti-drug-resistance agent.
  • Demonstrates superior antimicrobial activity against various clinical isolates.
  • Shows significant efficacy in *M. tuberculosis* infection models.
  • Reduces bacterial burden in *Staphylococcus aureus* septicemia models.
Catalog No. 50-004-13215 Supplier Medchemexpress LLC Supplier No. HYB1329500MG
Apramycin sulfate is an orally active, acidic pH tolerant aminoglycoside antibiotic. It inhibits protein biosynthesis by targeting bacterial ribosomes and is considered a potential anti-drug-resistance antibiotic. This compound demonstrates significant efficacy against various bacterial strains, including *K. pneumoniae*, *Enterobacter*, and *E. coli*, often showing better antimicrobial activity than other common antibiotics.
  • Orally active and acidic pH tolerant.
  • Tolerant to aminoglycoside-modifying enzymes.
  • Inhibits protein biosynthesis in bacteria.
  • Potential anti-drug-resistance agent.
  • Demonstrates superior antimicrobial activity against various clinical isolates.
  • Shows significant efficacy in *M. tuberculosis* infection models.
  • Reduces bacterial burden in *Staphylococcus aureus* septicemia models.