Learn More
Medchemexpress LLC Apramycin (sulfate) | 65710-07-8 | MFCD06200257 | 98% | 1 G
SDP
Catalog No. 5000413238
Encompass_Preferred
Apramycin (EBL 1003) sulfate is an orally active, acidic pH tolerant and aminoglycoside-modifying-enzymes-tolerant aminoglycoside antibiotic which inhibits protein biosynthesis by targeting the bacterial ribosome. Apramycin sulfate is a potential anti-drug-resistance antibiotic.
  • Orally active.
  • Acidic pH tolerant.
  • Aminoglycoside-modifying-enzymes-tolerant.
  • Inhibits protein biosynthesis by targeting the bacterial ribosome.
  • Potential anti-drug-resistance antibiotic.
  • Store at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.
Catalog No. 50-004-13238 Supplier Medchemexpress LLC Supplier No. HYB13291G
Apramycin (EBL 1003) sulfate is an orally active, acidic pH tolerant and aminoglycoside-modifying-enzymes-tolerant aminoglycoside antibiotic which inhibits protein biosynthesis by targeting the bacterial ribosome. Apramycin sulfate is a potential anti-drug-resistance antibiotic.
  • Orally active.
  • Acidic pH tolerant.
  • Aminoglycoside-modifying-enzymes-tolerant.
  • Inhibits protein biosynthesis by targeting the bacterial ribosome.
  • Potential anti-drug-resistance antibiotic.
  • Store at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.