Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apramycin Sulfate, G-Biosciences
SCI_ED

Catalog No. S162507
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162507 1 g
S157223 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162507 Supplier G-Biosciences Supplier No. RC888
Add to Cart
Add to Cart

Apramycin is an aminoglycoside antibiotic useful against gram-negative bacteria such as E. coli and Salmonella species.

It is used to study antibiotic resistance and protein synthesis translocation inhibition.

Chemical Identifiers

Molecular Weight (g/mol) 637.66 g/mol

Specifications

Boiling Point 823°C
Quantity 1 g
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.