Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apramycin sulfate

Catalog No. AAJ6661606
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6661606 5 g
AAJ6661603 1 g
AAJ6661609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6661606 Supplier Thermo Scientific Chemicals Supplier No. J6661606
Add to Cart

Apramycin sulfate, CAS # 65710-07-8, is the salt of apramycin, the bacteriostatic aminoglycoside antibiotic. | CAS: 65710-07-8 | C21H43N5O15S | 637.66 g/mol

Apramycin sulfate binds to the deep groove of RNA and effectively inhibits ribosomal translocation prohibiting protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Apramycin is a 2-deoxysteptamine derivative and an aminoglycoside originally isolated from Streptomyces tenebrarius
• Apramycin sulfate is comprised of a bicyclic sugar moiety and a monosubstituted deoxystreptamine attached to a sulfate group

Application

• Apramycin sulfate exhibits antibiotic activity against gram-negative bacteria
• It is suitable for laboratory use against Escherichia coli, Klebsiella pneumoniae, and Pseudomonas aeruginosa
• It is also used for laboratory study of infectious disease

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65710-07-8
Molecular Formula C21H43N5O15S
Molecular Weight (g/mol) 637.66
MDL Number MFCD06200257
InChI Key WGLYHYWDYPSNPF-RQFIXDHTSA-N
Synonym Nebramycin II
PubChem CID 3081544
IUPAC Name (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O

Specifications

Type Apramycin Sulfate
Physical Form Powder
Melting Point 168°C
Color Yellow
Quantity 5 g
Flash Point >110°C (230°F)
Boiling Point 823°C
Solubility Information Soluble in water at 25mg/ml
Formula Weight 637.65
Chemical Name or Material Apramycin sulfate
Hazard Category H315-H319-H360
Hazard Statement H360-H315-H319; May damage fertility or the unborn child. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 265-890-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.