Learn More
Cayman Chemical Apramycin (sulfate hydrate), Purity: ≥95%, 1 g
SDP
Catalog No. 502983319
Encompass_Preferred
An aminoglycoside antibiotic that is active against a variety of Gram-positive and Gram-negative bacteria, including strains which produce aminoglycoside-modifying enzymes
Catalog No. 50-298-3319 Supplier Cayman Chemical Supplier No. 196881 G
An aminoglycoside antibiotic that is active against a variety of Gram-positive and Gram-negative bacteria, including strains which produce aminoglycoside-modifying enzymes