Learn More
Medchemexpress LLC HY-12085 10mg , Apremilast CAS:608141-41-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1208510MG
Encompass_Preferred
HY-12085 10mg Apremilast CAS: 608141-41-9 Apremilast (CC-10004) is an orally available inhibitor of type-4 cyclic nucleotide phosphodiesterase (PDE-4) with an IC50 of 74 nM. Apremilast inhibits TNF-α release by lipopolysaccharide (LPS) with an IC50 of 104 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0603