Learn More
Abcam Apremilast, 25MG
Catalog No. 502671253
Encompass_Preferred
MW 460.5 Da, Purity >98%. Ppotent PDE4 (phosphodiesterase 4) inhibitor (IC₅₀ = 74 nM). It reduces TNF-α production in human synovial cells and significantly suppresses experimental arthritis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1253 Supplier Abcam Supplier No. AB28537925MG
MW 460.5 Da, Purity >98%. Ppotent PDE4 (phosphodiesterase 4) inhibitor (IC₅₀ = 74 nM). It reduces TNF-α production in human synovial cells and significantly suppresses experimental arthritis.
The product is subject to the following: Abcam Restricted Use Statement