Learn More
Selleck Chemical LLC Apremilast
SDP
Catalog No. 5000004254
Encompass_Preferred
Apremilast (CC-10004) is a potent and orally active PDE4 and TNF- inhibitor with IC50 of 74 nM and 77 nM respectively
Catalog No. 50-000-04254 Supplier Selleck Chemical LLC Supplier No. S80345MG
Apremilast (CC-10004) is a potent and orally active PDE4 and TNF- inhibitor with IC50 of 74 nM and 77 nM respectively