Learn More
Apexbio Technology LLC Apremilast (CC-10004), 25mg. Cas: 608141-41-9 MFCD: MFCD18782607. PDE4 inhibitor
SDP
Supplier:  Apexbio Technology LLC A431725
Encompass_Preferred
Apremilast, also known as CC-10004, is a novel and potent small-molecule inhibitor of phosphodiesterase 4 (PDE4), a key enzyme involved in cyclic adenosine monophosphate (cAMP) degradation and cytokine production of inflammatory cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4918