Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apremilast-d5 | 1258597-47-5 | 99.3% | 10 MG
SDP

Catalog No. 5000231133
Encompass_Preferred

Apremilast-d5 is a deuterium-labeled analogue of apremilast supplied as a purified analytical standard for use as a tracer or internal standard in quantitative methods. It is provided as a 10 mg powder and is intended for research use only.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity suitable for analytical reference use.
  • Supplied as a 10 mg powder with storage recommendations for long-term stability.
  • Molecular weight 465.53 g/mol and formula C22H19D5N2O7S.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-31133 Supplier Medchemexpress LLC Supplier No. HY12085S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apremilast-d5 is a deuterium-labeled analogue of apremilast supplied as a purified analytical standard for use as a tracer or internal standard in quantitative methods. It is provided as a 10 mg powder and is intended for research use only.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity suitable for analytical reference use.
  • Supplied as a 10 mg powder with storage recommendations for long-term stability.
  • Molecular weight 465.53 g/mol and formula C22H19D5N2O7S.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.