Learn More
Medchemexpress LLC Apremilast-d5 | 1258597-47-5 | 99.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12085S5MG
Encompass_Preferred
Apremilast-d5 is a deuterium-labeled analogue of apremilast provided as a high-purity solid for research use. It is intended for use as a tracer or internal standard in quantitative analytical methods and retains the pharmacological profile of apremilast as a PDE-4 inhibitor.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High purity supports analytical applications.
  • Supplied as a solid in milligram-scale quantities.
  • Retains PDE-4 inhibitory properties for research studies.
Catalog No. 50-002-32752