Learn More
Medchemexpress LLC Aprepitant | 170729-80-3 | MFCD08277635 | 534.43 g/mol | C23H21F7N4O3 | 1 ML
SDP
Catalog No. 5000214912
Encompass_Preferred
Aprepitant is a selective, high-affinity neurokinin 1 (NK1) receptor antagonist (Kd ≈ 86 pM) provided as a research-grade reagent for in vitro pharmacology and analytical applications. For research use only.
  • Selective neurokinin 1 (NK1) receptor antagonist.
  • High-affinity binding with Kd ≈ 86 pM.
  • Supplied as a 10 mM solution for convenient in vitro dosing.
  • Suitable for pharmacology, signaling, and analytical studies.
  • Well-characterized chemical identity (CAS 170729-80-3, MW 534.43 g/mol).
Catalog No. 50-002-14912 Supplier Medchemexpress LLC Supplier No. HY1005210MM 1ML
Aprepitant is a selective, high-affinity neurokinin 1 (NK1) receptor antagonist (Kd ≈ 86 pM) provided as a research-grade reagent for in vitro pharmacology and analytical applications. For research use only.
  • Selective neurokinin 1 (NK1) receptor antagonist.
  • High-affinity binding with Kd ≈ 86 pM.
  • Supplied as a 10 mM solution for convenient in vitro dosing.
  • Suitable for pharmacology, signaling, and analytical studies.
  • Well-characterized chemical identity (CAS 170729-80-3, MW 534.43 g/mol).