Learn More
Medchemexpress LLC Aprepitant | 170729-80-3 | MFCD08277635 | 99.8% | 534.43 g/mol | C23H21F7N4O3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10052200MG
Encompass_Preferred
Aprepitant is a selective, high-affinity neurokinin 1 (NK1) receptor antagonist used in research to probe NK1-mediated signaling and substance P pathways. It is supplied as a white to off-white solid with high stated purity and is suitable for analytical, pharmacological, and GPCR studies.
  • Selective neurokinin 1 receptor antagonist.
  • High affinity with a reported Kd of ~86 pM.
  • High purity suitable for analytical applications.
  • White to off-white solid appearance.
  • Available as powder and as DMSO solutions for convenience.
Catalog No. 50-002-14733