Learn More
Apexbio Technology LLC Aprepitant 170729-80-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063094
Encompass_Preferred
Aprepitant (MK-0869) is a highly selective antagonist of the neurokinin-1 (NK-1) receptor functioning by competitively inhibiting substance P (SP) an undecapeptide belonging to the tachykinin family from activating NK-1 receptors Its binding affinity for human NK-1 receptors is indicated by a dissociation constant (Kd) of approximately 86 pM In cellular models overexpressing NK-1 receptors including glioma (GAMG) neuroblastoma (SKN-BE2 IC50 19 6 M IMR-32 IC50 27 7 M KELLY IC50 30 4 M) retinoblastoma (Y-79 IC50 23 M WERI-Rb-1 IC50 31 2 M) pancreatic cancer (PA-TU-8902 IC50 27 4 M CAPAN-1 IC50 22 7 M) and colon cancer (SW-403 IC50 30 5 M) aprepitant application has been reported to inhibit tumor cell proliferation in vitro
Catalog No. 50-000-63094 Supplier Apexbio Technology LLC Supplier No. A16845.1
 Restricted Item
Add to Cart
Add to Cart
Aprepitant (MK-0869) is a highly selective antagonist of the neurokinin-1 (NK-1) receptor functioning by competitively inhibiting substance P (SP) an undecapeptide belonging to the tachykinin family from activating NK-1 receptors Its binding affinity for human NK-1 receptors is indicated by a dissociation constant (Kd) of approximately 86 pM In cellular models overexpressing NK-1 receptors including glioma (GAMG) neuroblastoma (SKN-BE2 IC50 19 6 M IMR-32 IC50 27 7 M KELLY IC50 30 4 M) retinoblastoma (Y-79 IC50 23 M WERI-Rb-1 IC50 31 2 M) pancreatic cancer (PA-TU-8902 IC50 27 4 M CAPAN-1 IC50 22 7 M) and colon cancer (SW-403 IC50 30 5 M) aprepitant application has been reported to inhibit tumor cell proliferation in vitro