Learn More
TARGETMOL CHEMICALS INC Aprepitant 200MG
SDP
Catalog No. 502878545
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Aprepitant is a Substance P/Neurokinin-1 Receptor Antagonist. The mechanism of action of aprepitant is as a Neurokinin 1 Antagonist, and Cytochrome P450 3A4 Inhibitor, and Cytochrome P450 2C9 Inducer, and Cytochrome P450 3A4 Inducer. Purity 99.9%
Catalog No. 50-287-8545 Supplier TARGETMOL CHEMICALS INC Supplier No. T1743200MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Aprepitant is a Substance P/Neurokinin-1 Receptor Antagonist. The mechanism of action of aprepitant is as a Neurokinin 1 Antagonist, and Cytochrome P450 3A4 Inhibitor, and Cytochrome P450 2C9 Inducer, and Cytochrome P450 3A4 Inducer. Purity 99.9%