Learn More
Apexbio Technology LLC Aprepitant, 5mg. Cas: 170729-80-3 MFCD: MFCD08277635. Substance P (SP) inhibitor
SDP
Supplier:  Apexbio Technology LLC A16845
Encompass_Preferred
Aprepitant (also known as MK-0869) is a novel and highly selective Neurokinin-1 (NK-1) receptor antagonist that inhibits the activity of substance P (SP), an undecapeptide belonging to the tachykinin family of peptides. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4704