Learn More
Medchemexpress LLC Aprepitant-d4 5mg | 1133387-60-6 | 538.45 | C23H17D4F7N4O3 | 5 MG
SDP
Catalog No. 5000230078
Encompass_Preferred
Aprepitant-d4 is the deuterium-labeled analogue of aprepitant, supplied as a powder and intended for use as a stable isotope internal standard in research and analytical laboratories, particularly for LC-MS quantitation.
  • Deuterium-labeled internal standard for quantitative LC-MS assays.
  • High chemical purity (99.0%).
  • Suitable molecular weight and isotopic composition for sensitive detection (MW 538.45).
  • Supplied as a stable powder with recommended storage at -20°C for long-term stability.
  • High solubility in DMSO (100 mg/mL); warming and sonication recommended.
Catalog No. 50-002-30078 Supplier Medchemexpress LLC Supplier No. HY1153975MG
Aprepitant-d4 is the deuterium-labeled analogue of aprepitant, supplied as a powder and intended for use as a stable isotope internal standard in research and analytical laboratories, particularly for LC-MS quantitation.
  • Deuterium-labeled internal standard for quantitative LC-MS assays.
  • High chemical purity (99.0%).
  • Suitable molecular weight and isotopic composition for sensitive detection (MW 538.45).
  • Supplied as a stable powder with recommended storage at -20°C for long-term stability.
  • High solubility in DMSO (100 mg/mL); warming and sonication recommended.