Learn More
Selleck Chemical LLC Aprepitant 5mg 170729-80-3 MK-0869, L-754030
SDP
Catalog No. 507617
Encompass_Preferred
Aprepitant (MK-0869, L-754030) is a potent and selective neurokinin-1 receptor antagonist with IC50 of 0.1 nM. Aprepitant reduces levels of pro-inflammatory cytokines including G-CSF, IL-6, IL-8 and TNF?. Aprepitant inhibits HIV infection of human macrophages. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-761-7 Supplier Selleck Chemical LLC Supplier No. S11895MG
Aprepitant (MK-0869, L-754030) is a potent and selective neurokinin-1 receptor antagonist with IC50 of 0.1 nM. Aprepitant reduces levels of pro-inflammatory cytokines including G-CSF, IL-6, IL-8 and TNF?. Aprepitant inhibits HIV infection of human macrophages. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.