Learn More
Cayman Chemical AprIcItabIn 10mg
SDP
Catalog No. 502911933
Encompass_Preferred
A nucleoside reverse transcriptase inhibitor; reduces HIV-1 infectivity in MT-4 and U937 cells (IC50s = 2.8 and 0.4 µM, respectively), as well as in isolated human CBMCs (IC50 = 0.4 µM)
Catalog No. 50-291-1933 Supplier Cayman Chemical Supplier No. 3603710 MG
A nucleoside reverse transcriptase inhibitor; reduces HIV-1 infectivity in MT-4 and U937 cells (IC50s = 2.8 and 0.4 µM, respectively), as well as in isolated human CBMCs (IC50 = 0.4 µM)