Learn More
Enterprise Technology Solutions Apricorn 10TB Aegis Padlock DT USB 3.0 External Desktop Drive
SDP
Supplier:  Enterprise Technology Solutions ADT3PL25610TB
Encompass_Preferred
The 10TB Aegis Padlock DT USB 3.0 External Desktop Drive from Apricorn is a secure desktop hard drive with on-the-fly 256-bit AES-XTS hardware encryption, PIN access, and a heat-dissipating aluminum enclosure.
Catalog No. 50-001-73229