Learn More
Enterprise Technology Solutions Apricorn 512GB Aegis Fortress USB 3.2 Gen 1 SSD w/ PIN Access
SDP
Supplier:  Enterprise Technology Solutions A253PL256S512F
Encompass_Preferred
The 512GB Aegis Fortress USB 3.2 Gen 1 SSD from Apricorn is certified to NIST FIPS 140-2 Level 2 requirements. This drive is tested on 11 cryptographic levels. All data on the drive is protected with real-time hardware AES-XTS 256-bit encryption.
Catalog No. 50-001-73279