Learn More
Enterprise Technology Solutions Apricorn Aegis Secure Key 3NX Encrypted USB 3.1 Gen 1 Flash Drive (16GB)
SDP
Catalog No. 5000173230
Encompass_Preferred
Securely stash away your sensitive data with the Aegis Secure Key 3NX Encrypted USB 3.1/3.0 Flash Drive (16GB) from Apricorn. Utilizing a USB Type-A connector, these drives feature full-disk 256-bit AES-XTS hardware encryption and are operated by a polymer-coated wear-resistant keypad built into the device. This keypad is designed to resist all key logging attempts.
Catalog No. 50-001-73230 Supplier Enterprise Technology Solutions Supplier No. ASK3NX16GB
Securely stash away your sensitive data with the Aegis Secure Key 3NX Encrypted USB 3.1/3.0 Flash Drive (16GB) from Apricorn. Utilizing a USB Type-A connector, these drives feature full-disk 256-bit AES-XTS hardware encryption and are operated by a polymer-coated wear-resistant keypad built into the device. This keypad is designed to resist all key logging attempts.