Learn More
Enzo Life Sciences APRIL polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADICSA836E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human APRIL. UniProt: O75888. Species Reactivity: Human, Mouse. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (10µg/ml). Western Blot (1-4µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-659-052