Learn More
AdipoGen APRIL (human) (H98) (multimeric) (rec.) MultiPack,
SDP
Supplier:  AdipoGen AG40B00883010
Encompass_Preferred
APRIL (human) (H98) (multimeric) (rec.) MultiPack, AdipoGen Life Sciences. Protein. The extracellular domain of human APRIL (aa 98-233) is fused at the N-terminus to human ACRP30headless (aa 16-108) and a FLAG¿-tag. Stimulates B cell proliferation. Lyophilized. Contains PBS + 0.5% Trehalose.
Catalog No. 50-117-5374