Learn More
Enzo Life Sciences APRIL (human) monoclonal antibody (Aprily-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804148C100
Encompass_Preferred
Clone: Aprily-1. Isotype: IgG1. Immunogen: Recombinant human APRIL (aa 93-233). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: Flow Cytometry, IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9795