Learn More
Enzo Life Sciences APRIL (human) monoclonal antibody (Aprily-8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804149C100
Encompass_Preferred
Clone: Aprily-8. Isotype: IgG1. Immunogen: Recombinant human APRIL (aa 93-233). Source: Purified from concentrated hybridoma tissue supernatant. Species reactivity: Human. Specificity: Recognizes APRIL and TWE-PRIL. Applications: Flow Cytometry, ICC, IHC (PS), IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9796