Learn More
Enzo Life Sciences APRIL-(human) monoclonal antibody (Aprily-5) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804801BC100
Encompass_Preferred
Clone: Aprily-5. Isotype: IgG1. Immunogen: Recombinant human APRIL (aa 93-233). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes full-length and cleaved APRIL. Applications: ELISA, IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9971