Learn More
Enzo Life Sciences APRIL-(membrane form) (human) polyclonal antibody (AT125) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210950C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human APRIL (membrane form) (aa 50-100). UniProt ID: O75888. Source: Purified from rabbit serum. Species reactivity: Human. Specificity: Recognizes membrane-bound form of human APRIL. Does not recognize the soluble form of human APRIL. Applications: WB. Recommended Dilutions: Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8192