Learn More
AdipoGen anti-APRIL (mouse), mAb (rec.) (blocking) (Apry-1-1) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG27B0001BC100
Encompass_Preferred
Recombinant Antibody. Recognizes mouse APRIL. Does not recognize mouse BAFF. Species cross-reactivity: Mouse. Clone: Apry-1-1. Isotype: Mouse IgG2blambda. Applications: ELISA, FUNC (Blocking), IP. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 0.02% sodium azide. APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumor cell growth and is involved in monocyte/macrophage-mediated immunological processes. Anti-APRIL (mouse) Monoclonal Antibody (recombinant) (Blocking) (APRY-1-1) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3012