Learn More
AdipoGen APRIL msH98multimeric
SDP
Supplier:  AdipoGen AG40B0035C010
Encompass_Preferred
Protein. The extracellular domain of mouse APRIL aa 98-232 is fused at the N-terminus to mouse ACRP30headless aa 18-110 and a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.5% Trehalose. Binds to human and mouse BCMA and TACI. Does not bind to proteoglycans. Purity >95% SDS-PAGE. APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumor cell growth, is involved in monocyte/macrophage-mediated immunological processes and functions as an important survival factor for plasmablasts and bone marrow plasma cells. MultimericAPRIL™ is a high activity construct in which two trimeric APRIL ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively stimulates proliferation B cell.
Catalog No. 50-256-2931