Learn More
Medchemexpress LLC APRIL/TNFSF13 Trimer 100ug
SDP
Catalog No. 5000197061
Encompass_Preferred
Recombinant human APRIL/TNFSF13 trimer produced in HEK293 cells and supplied with a His-Flag tag; designed for in vitro receptor binding and functional studies related to B-cell biology and lymphatic malignancies.
  • Suitable for receptor binding assays and biochemical characterization.
  • Trimeric form mimics the native ligand assembly for functional studies.
  • HEK293 expression supports mammalian folding and glycosylation.
  • Affinity tags enable straightforward purification and detection.
  • Optimized for cell signaling and ligand-receptor interaction experiments.
Catalog No. 50-001-97061 Supplier Medchemexpress LLC Supplier No. HYP70733A100UG
Add to Cart
Add to Cart
Recombinant human APRIL/TNFSF13 trimer produced in HEK293 cells and supplied with a His-Flag tag; designed for in vitro receptor binding and functional studies related to B-cell biology and lymphatic malignancies.
  • Suitable for receptor binding assays and biochemical characterization.
  • Trimeric form mimics the native ligand assembly for functional studies.
  • HEK293 expression supports mammalian folding and glycosylation.
  • Affinity tags enable straightforward purification and detection.
  • Optimized for cell signaling and ligand-receptor interaction experiments.