Learn More
Medchemexpress LLC APRIL/TNFSF13 Trimer 10ug
SDP
Catalog No. 5000196920
Encompass_Preferred
Recombinant human APRIL (TNFSF13) trimer produced in HEK293 cells and supplied lyophilized with N-terminal His and Flag tags. The protein is highly purified and low-endotoxin, formulated in buffered saline for stability, and intended for receptor binding, biochemical, or cell-based assays. Store frozen for long-term stability and aliquot to avoid repeated freeze-thaw cycles.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive biological assays.
  • Mammalian expression preserves native post-translational modifications.
  • N-terminal His and Flag tags facilitate detection and purification.
  • Lyophilized formulation enables convenient storage and transport.
  • Stable when aliquoted and frozen for extended storage.
Catalog No. 50-001-96920 Supplier Medchemexpress LLC Supplier No. HYP70733A10UG
Recombinant human APRIL (TNFSF13) trimer produced in HEK293 cells and supplied lyophilized with N-terminal His and Flag tags. The protein is highly purified and low-endotoxin, formulated in buffered saline for stability, and intended for receptor binding, biochemical, or cell-based assays. Store frozen for long-term stability and aliquot to avoid repeated freeze-thaw cycles.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive biological assays.
  • Mammalian expression preserves native post-translational modifications.
  • N-terminal His and Flag tags facilitate detection and purification.
  • Lyophilized formulation enables convenient storage and transport.
  • Stable when aliquoted and frozen for extended storage.