Learn More
Medchemexpress LLC APRIL/TNFSF13 Trimer 20ug | 20ug
SDP
Catalog No. 5000187202
Encompass_Preferred
APRIL protein (CD256) is a ligand in the tumor necrosis factor (TNF) family that can induce proliferation regulate tumor cell growth and may participate in mononuclear/macrophage-mediated immune process[1] APRIL protein is produced by myeloid cells and their precursors to accelerate cell maturation and peripheral rupture[2] APRIL protein has been widely used in the study of lymphatic malignancies APRIL protein is a type II membrane protein with cytoplasmic domain hydrophobic transmembrane domain and extracellular domain APRIL/TNFSF13 Trimer Protein Cynomolgus (HEK293 His-Flag-Avi) is produced by HEK293 cells (K112-L250) with N-terminal His Avi and Flag tags
Catalog No. 50-001-87202 Supplier Medchemexpress LLC Supplier No. HYP7756420UG
Add to Cart
Add to Cart
APRIL protein (CD256) is a ligand in the tumor necrosis factor (TNF) family that can induce proliferation regulate tumor cell growth and may participate in mononuclear/macrophage-mediated immune process[1] APRIL protein is produced by myeloid cells and their precursors to accelerate cell maturation and peripheral rupture[2] APRIL protein has been widely used in the study of lymphatic malignancies APRIL protein is a type II membrane protein with cytoplasmic domain hydrophobic transmembrane domain and extracellular domain APRIL/TNFSF13 Trimer Protein Cynomolgus (HEK293 His-Flag-Avi) is produced by HEK293 cells (K112-L250) with N-terminal His Avi and Flag tags