Learn More
Medchemexpress LLC April/TNFSF13 trimer protein, human (biotinylated) | >95.0% | 20UG
SDP
Catalog No. 5000177239
Encompass_Preferred
Biotinylated human APRIL (TNFSF13) trimer is a recombinant cytokine produced in HEK293 cells and supplied as a lyophilized powder with N-terminal Flag, 8xHis, and Avi tags. The product is glycosylated, has high purity and low endotoxin, and is intended for use in ELISA, binding, and receptor interaction assays.
  • HEK293-expressed, glycosylated trimeric human APRIL.
  • Biotinylated (Avi) with N-Flag and N-8xHis tags for detection and capture.
  • High purity (>95%) by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation with trehalose for stability and storage.
  • Validated in ELISA with EC50 ≤16.3 ng/mL for receptor binding.
Catalog No. 50-001-77239 Supplier Medchemexpress LLC Supplier No. HYP70066120UG
Biotinylated human APRIL (TNFSF13) trimer is a recombinant cytokine produced in HEK293 cells and supplied as a lyophilized powder with N-terminal Flag, 8xHis, and Avi tags. The product is glycosylated, has high purity and low endotoxin, and is intended for use in ELISA, binding, and receptor interaction assays.
  • HEK293-expressed, glycosylated trimeric human APRIL.
  • Biotinylated (Avi) with N-Flag and N-8xHis tags for detection and capture.
  • High purity (>95%) by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation with trehalose for stability and storage.
  • Validated in ELISA with EC50 ≤16.3 ng/mL for receptor binding.