Learn More
Medchemexpress LLC APRIL/TNFSF13 Trimer 50ug | 50ug
SDP
Catalog No. 5000186797
Encompass_Preferred
APRIL protein (CD256) is a ligand in the tumor necrosis factor (TNF) family that can induce proliferation regulate tumor cell growth and may participate in mononuclear/macrophage-mediated immune process[1] APRIL protein is produced by myeloid cells and their precursors to accelerate cell maturation and peripheral rupture[2] APRIL protein has been widely used in the study of lymphatic malignancies APRIL protein is a type II membrane protein with cytoplasmic domain hydrophobic transmembrane domain and extracellular domain APRIL/TNFSF13 Trimer Protein Cynomolgus (HEK293 His-Flag-Avi) is produced by HEK293 cells (K112-L250) with N-terminal His Avi and Flag tags
Catalog No. 50-001-86797 Supplier Medchemexpress LLC Supplier No. HYP7756450UG
APRIL protein (CD256) is a ligand in the tumor necrosis factor (TNF) family that can induce proliferation regulate tumor cell growth and may participate in mononuclear/macrophage-mediated immune process[1] APRIL protein is produced by myeloid cells and their precursors to accelerate cell maturation and peripheral rupture[2] APRIL protein has been widely used in the study of lymphatic malignancies APRIL protein is a type II membrane protein with cytoplasmic domain hydrophobic transmembrane domain and extracellular domain APRIL/TNFSF13 Trimer Protein Cynomolgus (HEK293 His-Flag-Avi) is produced by HEK293 cells (K112-L250) with N-terminal His Avi and Flag tags