Learn More
Medchemexpress LLC Aprindine hydrochloride | 33237-74-0 | 99.0% | 358.95 g·mol⁻¹ | C22H31ClN2 | 10 MG
SDP
Catalog No. 5000298784
Encompass_Preferred
Aprindine hydrochloride is a class I-b anti-arrhythmic research compound that blocks sodium channels (INa) and is used to study cardiac electrophysiology. Supplied as the hydrochloride salt in small milligram quantities, it is intended for analytical and preclinical research applications.
  • Class I-b anti-arrhythmic agent; sodium channel blocker.
  • Hydrochloride salt form for improved solubility.
  • Available in small, precise milligram pack sizes.
  • High purity suitable for analytical and preclinical studies.
  • Used in electrophysiology and cardiovascular research applications.
Catalog No. 50-002-98784 Supplier Medchemexpress LLC Supplier No. HYA0236A10MG
Aprindine hydrochloride is a class I-b anti-arrhythmic research compound that blocks sodium channels (INa) and is used to study cardiac electrophysiology. Supplied as the hydrochloride salt in small milligram quantities, it is intended for analytical and preclinical research applications.
  • Class I-b anti-arrhythmic agent; sodium channel blocker.
  • Hydrochloride salt form for improved solubility.
  • Available in small, precise milligram pack sizes.
  • High purity suitable for analytical and preclinical studies.
  • Used in electrophysiology and cardiovascular research applications.