Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aprindine hydrochloride | 33237-74-0 | 99.0% | C22H31ClN2 | 25 MG
SDP

Catalog No. 5000359165
Encompass_Preferred

Aprindine hydrochloride | 33237-74-0 | 99.0% | C22H31ClN2 | 25 MG

Catalog No. 50-003-59165 Supplier Medchemexpress LLC Supplier No. 25MGHYA0236A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aprindine hydrochloride | 33237-74-0 | 99.0% | C22H31ClN2 | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.