Learn More
Medchemexpress LLC Aprindine hydrochloride | 33237-74-0 | MFCD01698392 | 99.0% | C22H31ClN2 | 100 MG
SDP
Catalog No. 5000432387
Encompass_Preferred
Aprindine hydrochloride is an Ib-class anti-arrhythmic agent primarily used in the study of atrial fibrillation and ventricular arrhythmias. It functions by stabilizing cardiac electrical activity through multiple mechanisms.
  • Blocks sodium channels to reduce excitability and conduction velocity of cardiac muscle cells.
  • Inhibits delayed potassium currents, prolonging the atrial effective refractory period.
  • Inhibits atrial fibrillation.
  • Regulates intracellular calcium ion concentration by inhibiting the Na+/Ca2+ exchange current.
Catalog No. 50-004-32387 Supplier Medchemexpress LLC Supplier No. HYA0236A100MG
Aprindine hydrochloride is an Ib-class anti-arrhythmic agent primarily used in the study of atrial fibrillation and ventricular arrhythmias. It functions by stabilizing cardiac electrical activity through multiple mechanisms.
  • Blocks sodium channels to reduce excitability and conduction velocity of cardiac muscle cells.
  • Inhibits delayed potassium currents, prolonging the atrial effective refractory period.
  • Inhibits atrial fibrillation.
  • Regulates intracellular calcium ion concentration by inhibiting the Na+/Ca2+ exchange current.