Learn More
Medchemexpress LLC HY-15895 5mg Medchemexpress, Aprocitentan CAS:1103522-45-7 Purity:>98%
SDP
Catalog No. 502081821
Encompass_Preferred
Medchemexpress, HY-15895 5mg Aprocitentan CAS:1103522-45-7 Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1821 Supplier Medchemexpress LLC Supplier No. HY158955MG
Medchemexpress, HY-15895 5mg Aprocitentan CAS:1103522-45-7 Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.