Learn More
Medchemexpress LLC Aprocitentan-d4 | 1103522-45-7 | >98.0% | 550.22 g/mol | C16H10D4Br2N6O4S | 10 MG
SDP
Catalog No. 5000287966
Encompass_Preferred
Aprocitentan-d4 is a deuterium-labeled analogue of aprocitentan provided as a stable-isotope reference material for research and analytical applications. It is intended for use as an internal standard in quantitative mass spectrometry and for pharmacokinetic and metabolic studies where isotopically labeled references are required.
  • Deuterium-labeled (d4) analogue for mass spectrometry
  • Suitable as an internal standard for quantitative LC-MS/MS assays
  • Intended for research and analytical applications
  • Useful in pharmacokinetic and metabolic studies
  • Offered in small-mass quantities suitable for analytical workflows
Catalog No. 50-002-87966 Supplier Medchemexpress LLC Supplier No. HY15895S10MG
Aprocitentan-d4 is a deuterium-labeled analogue of aprocitentan provided as a stable-isotope reference material for research and analytical applications. It is intended for use as an internal standard in quantitative mass spectrometry and for pharmacokinetic and metabolic studies where isotopically labeled references are required.
  • Deuterium-labeled (d4) analogue for mass spectrometry
  • Suitable as an internal standard for quantitative LC-MS/MS assays
  • Intended for research and analytical applications
  • Useful in pharmacokinetic and metabolic studies
  • Offered in small-mass quantities suitable for analytical workflows