Learn More
Med Vet International X-Ray Apron w/ Quick Release - Large Burgundy w/ Black Paws, J0676ML
Catalog No. 5000495314
Encompass_Preferred
Shoulder pads included. Attractive maroon and black paw design. 0.5mm Pb equivalency. Quick release buckle, large 36"length
Catalog No. 50-004-95314 Supplier Med Vet International Supplier No. JOR676ML
Shoulder pads included. Attractive maroon and black paw design. 0.5mm Pb equivalency. Quick release buckle, large 36"length