Learn More
Med Vet International X-Ray Apron w/ Quick Release - Small Burgundy w/ Black Paws, J0676M
Catalog No. 5000495373
Encompass_Preferred
JorVet offers a wide range of x-ray aprons designed for safety but offering a colorful variety of fabric choices. X-Ray Aprons. Three different styles are offered. • Shoulder pads included • Attractive maroon and black paw design • 0.5mm Pb equivalency Qu
Catalog No. 50-004-95373 Supplier Med Vet International Supplier No. JOR676M
JorVet offers a wide range of x-ray aprons designed for safety but offering a colorful variety of fabric choices. X-Ray Aprons. Three different styles are offered. • Shoulder pads included • Attractive maroon and black paw design • 0.5mm Pb equivalency Qu