Description
- Tychem™ Thermopro for triple hazard protection
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Provides frontal protection
- For flash fire and electric arc hazards, partial body protective Tychem™ ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex™ thread
- Two buckle closure system
- Open back for wearer comfort.
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Gray color for discretionary needs
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Specifications
| Size | Small |
| Material | Tychem |
| Color | Orange |
| Closure Type | Waist Ties |
| Cuff Style | Elastic |
| Seam Type | Taped |