Learn More
Research Products International Corp Aprotinin Solution 10,000 KIU/mL, 10 ML
SDP
Supplier:  Research Products International Corp A2057510.0
Encompass_Preferred
Aprotinin is a competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein, and plasmin. Material is provided as a solution containing 10,000 KIU/mL of aprotinin.
Catalog No. 50-136-8049