Learn More
Research Products International Corp Aprotinin, Bovine Lung, 50 Milligrams
SDP
Catalog No. 50488491
Encompass_Preferred
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at a pH>10 or <3.2. Target enzumes include trypsin, chymatrypsin, kalikrein and plasmin.
Activity: >4500 KIU/mg
Catalog No. 50-488-491 Supplier Research Products International Corp Supplier No. A205500.05
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at a pH>10 or <3.2. Target enzumes include trypsin, chymatrypsin, kalikrein and plasmin.
Activity: >4500 KIU/mg