Learn More
Gold Biotechnology Inc Aprotinin 25 mg
SDP
Catalog No. 501532749
Encompass_Preferred
Aprotinin is a monomeric globular polypeptide derived from bovine lung tissue. It consists of a chain of 58 amino acids that fold into a stable, compact tertiary structure of the 'small SS-rich type
Catalog No. 50-153-2749 Supplier Gold Biotechnology Inc Supplier No. A65525
Aprotinin is a monomeric globular polypeptide derived from bovine lung tissue. It consists of a chain of 58 amino acids that fold into a stable, compact tertiary structure of the 'small SS-rich type