Learn More
Carbosynth LLC. Aprotinin, 25mg, Disulfide bonds: Cys5-Cys55, Cys14-Cys38, and Cys30-Cys51, MW: (M.W. 6511.57), Molecular formula: C284H432N84O79S7, CAS# [9087-70-1]
SDP
Supplier:  Carbosynth LLC. IAT3830PI25MG
Encompass_Preferred
Synonyms: Arg-Pro-Asp-Phe-Cys-Leu-Glu-Pro-Pro-Tyr-Thr-Gly-Pro-Cys-Lys-Ala-Arg-Ile-Ile-Arg-Tyr-Phe-Tyr-Asn-Ala-Lys-Ala-Gly-Leu-Cys-Gln-Thr-Phe-Val-Tyr-Gly-Gly-Cys-Arg-Ala-Lys-Arg-Asn-Asn-Phe-Lys-Ser-Ala-Glu-Asp-Cys-Met-Arg-Thr-Cys-Gly-Gly-Ala, Application: Inhibitor for Trypsin, u-PA, Papain, and Cathepsin A/B, Storage Temperature: -20°C, References: I. Trautschold, E. Werle and G. Zickgraf-Rudel, Biochem. Pharm., 16, 59 (1967).
Catalog No. 50-168-5824