Learn More
Bachem Aprotinin Acetate
SDP
Supplier:  Bachem 4035911.0005
Encompass_Preferred
Aprotinin Acetate, H-Arg-Pro-Asp-Phe-Cys-Leu-Glu-Pro-Pro-Tyr-Thr-Gly-Pro-Cys-Lys-Ala-Arg-Ile-Ile-Arg-Tyr-Phe-Tyr-Asn-Ala-Lys-Ala-Gly-Leu-Cys-Gln-Thr-Phe-Val-Tyr-Gly-Gly-Cys-Arg-Ala-Lys-Arg-Asn-Asn-Phe-Lys-Ser-Ala-Glu-Asp-Cys-Met-Arg-Thr-Cys-Gly-Gly-Ala-OH, 9087-70-1, C284H432N84O79S7, Mr 6511.53, 5mg. Basic Protease Inhibitor, Pancreatic Trypsin Inhibitor, BPI, BPTI. Aprotinin (BPTI), a widely used serine protease inhibitor. Bachem's aprotinin is a synthetic product and is not derived from bovine origin.
Catalog No. NC1542892